Product Name
3-Chloro-2,4,5-trifluorobenzoic acid 97%
Product Number
643300
Product Brand
ALDRICH
CAS Number
101513-77-3
Molecular Weight
210.54
Test
Specification
APPEARANCEWHITE TO TAN POWDER, CRYSTALS, CRYSTALLINE
 POWDER AND/OR CHUNKS
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
TITRATION96.5%-103.5% (WITH NAOH)
GAS LIQUID96.5% (MINIMUM)
 APPROVED APRIL 10, 2007 RJM