Product Name
(R)-C3-TunePhos
Product Number
650862
Product Brand
ALDRICH
CAS Number
301847-89-2
Molecular Weight
594.62
Test
Specification
APPEARANCEWHITE TO OFF-WHITE POWDER, CRYSTALS OR
 CRYSTALLINE POWDER AND/OR CHUNKS
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
ELEMENTAL ANALYSISCARBON 69.7% - 88.6%
OPTICAL ROTATION-252 DEGREES +/- 5 DEGREES (C=1%, CHCL3)
 APPROVED MARCH 14, 2005 BTR