Product Name
4-Fluorophenethyl bromide 97%
Product Number
652008
Product Brand
ALDRICH
CAS Number
332-42-3
Molecular Weight
203.05
Test
Specification
Appearance (Color)Colorless to Yellow
Appearance (Form)Liquid
Infrared spectrumConforms to Structure
Purity (GC)≥96.5 %
 EXCLUDING TMCS