Product Name
Ethyl 2-amino-4-(4-bromophenyl)thiazole-5-carboxylate 90%
Product Number
653500
Product Brand
ALDRICH
CAS Number
834885-05-1
Molecular Weight
327.20
Test
Specification
APPEARANCEWHITE TO PINK TO TAN POWDER, CRYSTALS,
 FLAKES, CRYSTALLINE POWDER AND/OR CHUNKS
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
HIGH PRESSURE LIQUID88.0% (MINIMUM)
 APPROVED JANUARY 18, 2007 BTR