Product Name
5-Nitro-2-oxindole 97%
Product Number
653683
Product Brand
ALDRICH
CAS Number
20870-79-5
Molecular Weight
178.14
Test
Specification
Appearance (Color)Conforms to Requirements
 White to Tan to Light Orange
Appearance (Form)Conforms to Requirements
 Powder, Crystals, Crystalline Powder and/or Chunks
Infrared spectrumConforms to Structure
Carbon52.0 - 55.8 %
Nitrogen15.1 - 16.3 %
Purity (HPLC)≥96.5 %