Product Name
5-Bromo-2-(trifluoromethyl)pyridine 97%
Product Number
661104
Product Brand
ALDRICH
CAS Number
436799-32-5
Molecular Weight
225.99
Test
Specification
APPEARANCE (COLOR)White to Brown
APPEARANCE (FORM)Powder or Crystals
TITRATION96.5 - 103.5 %
TITRATION (METHOD)WITH AGNO3 AFTER O2 COMBUSTION
PURITY (GC AREA %)≥ 96.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE