Product Name
(1S,2S,4R)-(+)-Limonene-1,2-diol ≥97.0% (GC)
Product Number
669768
Product Brand
ALDRICH
CAS Number
38630-75-0
Molecular Weight
170.25
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals or Powder with Chunk(s) or Crystals with Chunk(s)
PURITY (GC AREA %)≥ 97.0 %
SPECIFIC ROTATION (20/D)45 ± 5 Degrees
CONCENTRATIONC=1 IN ACETONE
CARBON CONTENT70.55 % (THEORY)
HYDROGEN CONTENT10.66 % (THEORY)
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD60 MONTHS