Product Name
cataCXium® A 95%
Product Number
671479
Product Brand
ALDRICH
CAS Number
321921-71-5
Molecular Weight
358.54
Test
Specification
APPEARANCE (COLOR)White to Dark Yellow and Faint Beige to Dark Beige
APPEARANCE (FORM)Powder or Crystals
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
PHOSPHORUS 31 NMR SPECTRUMCONFORMS TO STRUCTURE
REMARKSChemical Purity >= 95 %
RECOMMENDED RETEST PERIOD40 MONTHS