Product Name
Neodymium-iron-boron, alloy 30/200 disc, size 13 mm × 6 mm
Product Number
693820
Product Brand
ALDRICH
CAS Number
Molecular Weight
Test
Specification
APPEARANCEWHITE TO GREY DISCS
ICP ASSAYWT% ~ 29% ND, 1.1% B, 0.8% AL, 0.5% NB
 3.6% DY, REMAINDER FE
VENDOR INFORMATIONMAX ENERGY PROUDCT, BH: ~250 KJ/M3
 RESIDUEAL INDUCTION, BR: ~1.14 T
 COERSIVE FORCE, HC: ~835 KA/M
 INT COERSIVE FORCE, HCI: ~2400 KA/M
 MAXIMUM TEMPERATURE RATING: 200 �C
 APPROVED JANUARY 22, 2008 LUKE GROCHOLL