Product Name
5-Hexylthiophene-2-carboxaldehyde 97%
Product Number
699187
Product Brand
ALDRICH
CAS Number
100943-46-2
Molecular Weight
196.31
Test
Specification
APPEARANCECOLORLESS TO YELLOW TO TAN LIQUID
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
TITRATION96.5-103.5% (WITH HYDROXYLAMINE)
GAS LIQUID96.5% (MINIMUM)
 APPROVED APRIL 14, 2008 DJP