Product Name
5-Bromobenzoxazole-2-thiol 97%
Product Number
699977
Product Brand
ALDRICH
CAS Number
439607-87-1
Molecular Weight
230.08
Test
Specification
APPEARANCEYELLOW TO BROWN POWDER OR CRYSTALS
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
TITRATION96.5-103.5% (WITH AGNO3 AFTER O2 COMBUSTION)
 AND/OR
ELEMENTAL ANALYSISCARBON 35.3% - 37.8%
 NITROGEN 5.8% - 6.4%
 SULFUR 13.4% - 14.5%
HIGH PRESSURE LIQUID96.5% (MINIMUM)
 APPROVED AUGUST 6, 2008 BTR