Product Name
2,3-Dibromo-4-methylthiophene 97%
Product Number
700282
Product Brand
ALDRICH
CAS Number
125257-38-7
Molecular Weight
255.96
Test
Specification
Appearance (Form)Liquid
Infrared spectrumConforms to Structure
Titration by AgNO396.5 - 103.5 %
 After Oxygen Combustion
Purity (GC)≥96.5 %