Product Name
3-Methyl-2-benzoxazolinone 98%
Product Number
711594
Product Brand
ALDRICH
CAS Number
21892-80-8
Molecular Weight
149.15
Test
Specification
APPEARANCE (COLOR)White to Light Brown to Light Red
APPEARANCE (FORM)Powder or Crystals or Needles
PURITY (GC AREA %)≥ 97.5 %
MELTING POINT83 - 87 C
CARBON CONTENT62.8 - 66.0 %
NITROGEN CONTENT9.2 - 9.6 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE