Product Name
(Iodomethyl)cyclopropane ≥95.0% (GC)
Product Number
713457
Product Brand
ALDRICH
CAS Number
33574-02-6
Molecular Weight
182.00
Test
Specification
APPEARANCE (COLOR)Very Faint Yellow to Very Dark Yellow
APPEARANCE (FORM)Liquid
TITRATION96.0 - 104.0 %
TITRATION (METHOD)ARGENTOMETRIC AFTER OXYGEN COMBUSTION
PURITY (GC AREA %)≥ 95.0 %
REFRACTIVE INDEX N20/D1.538 - 1.546
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE