Product Name
Sodium L-lactate ≥99.0% (NT)
Product Number
71718
Product Brand
ALDRICH
CAS Number
867-56-1
Molecular Weight
112.06
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder or Crystals
TITRATION (NT) HCLO4 0.1M99.0 - 101.0 %
ENANTIOMERIC EXCESS≥ 97.0 % (enzymatic)
SPECIFIC ROTATION (20/D)-13.0 - -12.0 Degrees
CONCENTRATIONC=1 IN WATER
WATER≤ 1.0 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD36 MONTHS