Product Name
Heptadecafluorooctanesulfonic acid potassium salt ≥98.0% (T)
Product Number
77282
Product Brand
ALDRICH
CAS Number
2795-39-3
Molecular Weight
538.22
Test
Specification
APPEARANCE (COLOR)White to Faint Beige
APPEARANCE (FORM)Powder or Crystals
TITRATION (ION EXCHANGE)≥ 98.0 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE