Product Name
1-Azido-2-fluorobenzene solution 0.5 M in tert-butyl methyl ether, ≥95% (HPLC)
Product Number
778842
Product Brand
ALDRICH
CAS Number
3296-04-6
Molecular Weight
137.11
Test
Specification
APPEARANCE (COLOR)Colorless to Very Light Yellow
APPEARANCE (FORM)Liquid
PURITY (HPLC AREA %)≥ 95.0 %
DENSITY D20/40.770 - 0.790
WATER (COULOMETR.)≤ 2.0 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
REMARKSCONCENTRATION ~0.5M IN TBME