Product Name
1-Azido-3-chlorobenzene solution 0.5 M in tert-butyl methyl ether, ≥97% (HPLC)
Product Number
779938
Product Brand
ALDRICH
CAS Number
3296-06-8
Molecular Weight
153.57
Test
Specification
APPEARANCE (COLOR)Colorless to Yellow and Colorless to Brown-Yellow
APPEARANCE (FORM)Liquid
PURITY (HPLC AREA %)≥ 97 %
WATER≤ 2.0 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
REMARKSCONCENTRATION ~0.5M IN TBME