Product Name
Sodium bis(trimethylsilyl)amide solution produced by Wacker Chemie AG, Burghausen, Germany, 40% in THF
Product Number
80631
Product Brand
ALDRICH
CAS Number
1070-89-9
Molecular Weight
183.37
Test
Specification
APPEARANCE (COLOR)Faint Yellow to Dark Yellow and Faint Brown-Yellow to Dark Brown-Yellow and Faint Brown to Dark Brown
APPEARANCE (FORM)Liquid
APPEARANCE (TURBIDITY)Clear
TITRATION40.0 - 42.0 %
TITRATABLE BASE≤ 0.5 % NAOH
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE