Product Name
Mowiol® 18-88 Mw ~130,000
Product Number
81365
Product Brand
ALDRICH
CAS Number
9002-89-5
Molecular Weight
Test
Specification
APPEARANCE (COLOR)White to Faint Beige
APPEARANCE (FORM)Crystals or Flakes
RESIDUE ON IGNITION≤ 0.50 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
VISCOSITY (ROTATION)16 - 20 mPas
VISCOSITY (CONDITIONS)4 % in H2O, 20 C
MISCELLANEOUS TESTSDEGREE OF HYDROLYSIS 86.7-88.7 % MOL