Product Name
Mowiol® 4-88 Mw ~31,000
Product Number
81381
Product Brand
ALDRICH
CAS Number
9002-89-5
Molecular Weight
Test
Specification
APPEARANCE (COLOR)White to Faint Yellow to Faint Beige
APPEARANCE (FORM)Crystals
RESIDUE ON IGNITION≤ 0.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
VISCOSITY3 - 5 MPAS (20 C, 4 % IN H2O)
MISCELLANEOUS TESTSDEGREE OF HYDROLYSIS 86.7-88.7 % MOL