Product Name
Mowiol® 4-98 Mw ~27,000
Product Number
81382
Product Brand
ALDRICH
CAS Number
9002-89-5
Molecular Weight
Test
Specification
APPEARANCE (COLOR)White to Light Yellow
APPEARANCE (FORM)Powder or Crystals or Flakes
RESIDUE ON IGNITION≤ 0.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
VISCOSITY (STOKES)4 - 5 mPas
VISCOSITY (CONDITIONS)20 C , 4 % IN H2O
MISCELLANEOUS TESTSDEGREE OF HYDROLYSIS 98.0-98.8 % MOL