Product Name
Silver perchlorate hydrate ≥96.0% (calc. based on dry substance, AT)
Product Number
85271
Product Brand
ALDRICH
CAS Number
331717-44-3
Molecular Weight
207.32 (anhydrous basis)
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder or Crystals
TITRATION96.0 - 103.0 % (in dried substance)
LOSS ON DRYING4.0 - 12.0 %