Product Name
Tetrabutylammonium acetate technical, ≥90% (T)
Product Number
86849
Product Brand
ALDRICH
CAS Number
10534-59-5
Molecular Weight
301.51
Test
Specification
APPEARANCE (COLOR)White to Faint Yellow
APPEARANCE (FORM)Powder or Crystals
TITRATION (ION EXCHANGE)90.0 - 105.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD42 MONTHS