Product Name
Tetrabutylammonium heptadecafluorooctanesulfonate ≥95.0% (H-NMR)
Product Number
86911
Product Brand
ALDRICH
CAS Number
111873-33-7
Molecular Weight
741.59
Test
Specification
APPEARANCE (COLOR)Yellow to Very Dark Yellow
APPEARANCE (FORM)Liquid or Viscous Liquid
INFRARED SPECTRUMCONFORMS TO STRUCTURE
ASSAY (QUANTITATIVE NMR)≥ 95.0 %
NMR-Internal StandardDimethylterephthalat