Product Name
2-Aminoheptane 99%
Product Number
A56205
Product Brand
ALDRICH
CAS Number
123-82-0
Molecular Weight
115.22
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE