Product Name
4-Aminosalicylic acid 99%
Product Number
A79604
Product Brand
ALDRICH
CAS Number
65-49-6
Molecular Weight
153.14
Test
Specification
APPEARANCE (COLOR)White to Beige
APPEARANCE (FORM)Powder
TITRATION (NT) HCLO4 0.1M98.5 - 101.5 %
PURITY (HPLC AREA %)≥ 98.5 %
SOLUBILITY (COLOR)Colorless to Faint Yellow and Colorless to Faint Brown-Yellow and Colorless to Faint Brown
SOLUBILITY (TURBIDITY)Clear to Slightly Hazy
SOLUBILITY (METHOD)50 MG in 1ML ETHANOL
INFRARED SPECTRUMCONFORMS TO STRUCTURE