Product Name
4,4′-Dimethoxybenzoin 95%
Product Number
A88409
Product Brand
ALDRICH
CAS Number
119-52-8
Molecular Weight
272.30
Test
Specification
APPEARANCE (COLOR)White to Yellow
APPEARANCE (FORM)Powder
PURITY (GC AREA %)≥ 94.5 %
PURITY (HPLC AREA %)≥ 94.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE