Product Name
3-Bromobenzaldehyde 97%
Product Number
B57206
Product Brand
ALDRICH
CAS Number
3132-99-8
Molecular Weight
185.02
Test
Specification
APPEARANCE (COLOR)Colorless to Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 96.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE