Product Name
(1S)-(+)-10-Camphorsulfonic acid 99%
Product Number
C2107
Product Brand
ALDRICH
CAS Number
3144-16-9
Molecular Weight
232.30
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
TITRATION (T) NAOH 0.1M98.5 - 101.5 %
OPTICAL ROTATION+22 +/- 2 DEGREES
CONCENTRATIONC=2%, H2O
SOLUBILITY (COLOR)Colorless to Faint Yellow
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)50MG/ML, H2O
INFRARED SPECTRUMCONFORMS TO STRUCTURE