Product Name
2,6-Diamino-4-chloropyrimidine 98%
Product Number
C33204
Product Brand
ALDRICH
CAS Number
156-83-2
Molecular Weight
144.56
Test
Specification
Appearance (Color)White to Tan
Appearance (Form)Conforms to Requirements
 Powder, Crystals, Crystalline Powder and/or Chunks
Infrared spectrumConforms to Structure
Purity (Titration by HClO4)97.5 - 102.5 %
Purity (GC)≥97.5 %