Product Name
3,5-Dimethyl-1-pyrazolylformaminidium nitrate 97%
Product Number
D182257
Product Brand
ALDRICH
CAS Number
38184-47-3
Molecular Weight
201.18
Test
Specification
APPEARANCE (COLOR)White to Beige
APPEARANCE (FORM)Powder
TITRATION (NT) HCLO4 0.1M96.5 - 103.5 %
PURITY (TLC AREA %)≥ 96.5 %
SOLUBILITY (COLOR)Colorless to Faint Yellow
SOLUBILITY (TURBIDITY)Clear to Very Slightly Hazy
SOLUBILITY (METHOD)100 MG/ML, WATER
INFRARED SPECTRUMCONFORMS TO STRUCTURE