Product Name
1,3-Dichloropropane 99%
Product Number
D72204
Product Brand
ALDRICH
CAS Number
142-28-9
Molecular Weight
112.99
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE