Product Name
2-Aminoethyl diphenylborinate 97%
Product Number
D9754
Product Brand
ALDRICH
CAS Number
524-95-8
Molecular Weight
225.09
Test
Specification
APPEARANCE (COLOR)White to Light Yellow and Faint Beige to Light Beige and Faint Brown to Light Brown
APPEARANCE (FORM)Powder
TITRATION (NT) HCLO4 0.1M96.5 - 103.5 %
SOLUBILITY (COLOR)Colorless to Light Brown
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)100 MG/ML IN METHANOL
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE