Product Name
Ethyl 2-benzylacetoacetate 97%
Product Number
E13202
Product Brand
ALDRICH
CAS Number
620-79-1
Molecular Weight
220.26
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 96.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE