Product Name
9-Hydroxyfluorene 96%
Product Number
H31204
Product Brand
ALDRICH
CAS Number
1689-64-1
Molecular Weight
182.22
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Crystal or Chunk(s)
PURITY (HPLC AREA %)≥ 95.5 %
CARBON CONTENT81.8 - 90.4 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE