Product Name
Iodoacetamide 97%
Product Number
I6709
Product Brand
ALDRICH
CAS Number
144-48-9
Molecular Weight
184.96
Test
Specification
APPEARANCEWHITE TO BROWN CYRSTALS OR POWDER
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
ELEMENTAL ANALYSISCARBON 12.5% - 13.4%
 NITROGEN 7.3% - 7.8%
 AND/OR
TITRATION96.5% - 103.5% (WITH AGNO3 AFTER OXYGEN
 COMBUSTION)
THIN-LAYERCONSISTENT WITH 97% PURITY
SOLUBILITY10% IN H2O; CLEAR, COLORLESS TO YELLOW
 SOLUTION
 REVISED NOVEMBER 12, 2002 JSB