Product Name
2,3-Lutidine 99%
Product Number
L3501
Product Brand
ALDRICH
CAS Number
583-61-9
Molecular Weight
107.15
Test
Specification
APPEARANCE (COLOR)Colorless to Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE