Product Name
(S)-(+)-Mandelic acid ReagentPlus®, ≥99%
Product Number
M2004
Product Brand
ALDRICH
CAS Number
17199-29-0
Molecular Weight
152.15
Test
Specification
APPEARANCEWHITE CRYSTALS
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
TITRATION99.0%-101.0% (WITH NAOH)
OPTICAL ROTATION+154 +/- 2 DEGREES (C = 2.8%, H2O)
GAS LIQUID99.0% (MINIMUM)
ENANTIOMERIC EXCESS98.5% (MINIMUM)
 REVISED JANUARY 28, 2005 JSB