Product Name
4′-Methoxypropiophenone ≥99%
Product Number
M24809
Product Brand
ALDRICH
CAS Number
121-97-1
Molecular Weight
164.20
Test
Specification
APPEARANCE (COLOR)Colorless to Light Yellow and White to Light Yellow
APPEARANCE (FORM)Liquid or Solid
PURITY (GC AREA %)≥ 99.0 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE