Product Name
3-Methylpyrazole 97%
Product Number
M75802
Product Brand
ALDRICH
CAS Number
1453-58-3
Molecular Weight
82.10
Test
Specification
APPEARANCE (COLOR)Colorless to Yellow to Brown-Yellow
APPEARANCE (FORM)Liquid
REM. ON APPEARANCEMATERIAL DARKENS IN STORAGE WITH NO LOSS OF PURITY
PURITY (GC AREA %)≥ 96.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE