Product Name
Octadecanamide technical grade, 85%
Product Number
O601
Product Brand
ALDRICH
CAS Number
124-26-5
Molecular Weight
283.49
Test
Specification
Appearance (Color)White to Beige
Appearance (Form)Conforms to Requirements
 Powder, Flakes, Chips or Beads
Infrared spectrumConforms to Structure
Purity (GC)≥84.0 %
Carbon64.0 - 88.4 %
Nitrogen4.1 - 5.7 %