Product Name
4-Phenylbutyric acid 99%
Product Number
P21005
Product Brand
ALDRICH
CAS Number
1821-12-1
Molecular Weight
164.20
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystal or Chunk(s)
TITRATION (T) NAOH 0.1M98.5 - 101.5 %
PURITY (GC AREA %)≥ 98.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE