Product Name
Hexylbenzene 97%
Product Number
P25701
Product Brand
ALDRICH
CAS Number
1077-16-3
Molecular Weight
162.27
Test
Specification
Appearance (Color)Colorless
Appearance (Form)Liquid
Infrared spectrumConforms to Structure
Purity (GC)≥96.5 %