Product Name
4-Phenyltoluene 98%
Product Number
P36606
Product Brand
ALDRICH
CAS Number
644-08-6
Molecular Weight
168.23
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder
PURITY (GC AREA %)≥ 97.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE