Product Name
Potassium hexahydroxoantimonate(V)
Product Number
P4308
Product Brand
ALDRICH
CAS Number
12208-13-8
Molecular Weight
262.90
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder
REDOX TITRATION44.3 - 49.8 %
REDOXTITRATION (METHOD)ANTIMONY CONTENT BY IODINE TITRATION
LOSS ON DRYING≤ 10.0 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE