Product Name
2-Pyridinealdoxime methiodide 99%
Product Number
P60205
Product Brand
ALDRICH
CAS Number
94-63-3
Molecular Weight
266.08
Test
Specification
APPEARANCEYELLOW CRYSTALS OR POWDER
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
ELEMENTAL ANALYSISCARBON 31.4%-32.3%
 NITROGEN 10.4%-10.8%
THIN-LAYERCONSISTENT WITH 99% PURITY
SOLUBILITY50MG/ML, H2O; CLEAR, YELLOW TO YELLOW-GREEN
 SOLUTION
 APPROVED JANUARY 9, 2009 RJM