Product Name
Trimethylene oxide 97%
Product Number
T76201
Product Brand
ALDRICH
CAS Number
503-30-0
Molecular Weight
58.08
Test
Specification
APPEARANCECOLORLESS LIQUID
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
GAS LIQUID96.5% (MINIMUM)
 APPROVED JULY 26, 2006 RJM