Product Name
Ursolic acid ≥90%
Product Number
U6753
Product Brand
ALDRICH
CAS Number
77-52-1
Molecular Weight
456.70
Test
Specification
Appearance (Color)White to Off-White
Appearance (Form)Powder
Solubility (Color)Colorless to Faint Yellow
Solubility (Turbidity)Clear to Slightly Hazy
 at 50 mg plus 1 mL of Dimethylformamide
Purity (GC)≥90 %