Product Name
Benzoin ≥98%
Product Number
W213209
Product Brand
ALDRICH
CAS Number
119-53-9
Molecular Weight
212.24
Test
Specification
APPEARANCEWHITE TO LIGHT YELLOW POWDER
MELTING POINT134 - 138 DEGREES CELSIUS
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
GAS LIQUID98.0% (MINIMUM)
SOLUBILITY50 MG/ML CHCL3; CLEAR TO SLIGHTLY HAZY,
 COLORLESS TO LIGHT YELLOW SOLUTION
 REVISED JULY 17, 2000 JSB